MilliporeSigma
  • Substituted 4-(2,2-diphenylethyl)pyridine-N-oxides as phosphodiesterase-4 inhibitors: SAR study directed toward the improvement of pharmacokinetic parameters.
Substituted 4-(2,2-diphenylethyl)pyridine-N-oxides as phosphodiesterase-4 inhibitors: SAR study directed toward the improvement of pharmacokinetic parameters.
Bioorganic & medicinal chemistry letters (2002-09-25)
Richard Frenette, Marc Blouin, Christine Brideau, Nathalie Chauret, Yves Ducharme, Richard W Friesen, Pierre Hamel, Tom R Jones, France Laliberté, Chun Li, Paul Masson, Malia McAuliffe, Yves Girard
ABSTRACT
A detailed SAR study directed toward the optimization of pharmacokinetic parameters for analogues of L-791,943 is reported. The introduction of a soft metabolic site on this structure permitted the identification of L-826,141 as a potent phosphodiesterase type 4 (PDE4) inhibitor that is well absorbed and that presents a shorter half-life than L-791,943 in a variety of animal species. The efficacy of L-826,141 is also demonstrated in different in vivo models.