Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

542660

Sigma-Aldrich

2-(Trifluoromethyl)benzenethiol

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4SH
CAS Number:
Molecular Weight:
178.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.499 (lit.)

bp

175-177 °C (lit.)

density

1.35 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)c1ccccc1S

InChI

1S/C7H5F3S/c8-7(9,10)5-3-1-2-4-6(5)11/h1-4,11H

InChI key

XVGQHRKNXSUPEF-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.9 °F - closed cup

Flash Point(C)

68.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel M Kroupa et al.
The journal of physical chemistry letters, 9(12), 3425-3433 (2018-06-03)
We studied the optical absorption enhancement in colloidal suspensions of PbS quantum dots (QD) upon ligand exchange from oleate to a series of cinnamate ligands. By combining experiments and ab initio simulations, we elucidate physical parameters that govern the optical
Carlos R Medrano et al.
The journal of physical chemistry letters, 9(12), 3517-3524 (2018-06-12)
For efficient conversion of light into useful energy sources, it is very important to study and describe the first steps of primary charge-transfer process in natural structures and artificial devices. The time scale of these processes in artificial photosynthetic and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service