MilliporeSigma
Optical Absorbance Enhancement in PbS QD/Cinnamate Ligand Complexes.
The journal of physical chemistry letters (2018-06-03)
Daniel M Kroupa, Márton Vörös, Nicholas P Brawand, Noah Bronstein, Brett W McNichols, Chloe V Castaneda, Arthur J Nozik, Alan Sellinger, Giulia Galli, Matthew C Beard
ABSTRACT
We studied the optical absorption enhancement in colloidal suspensions of PbS quantum dots (QD) upon ligand exchange from oleate to a series of cinnamate ligands. By combining experiments and ab initio simulations, we elucidate physical parameters that govern the optical absorption enhancement. We find that, within the cinnamate/PbS QD system, the optical absorption enhancement scales linearly with the electronic gap of the ligand, indicating that the ligand/QD coupling occurs equally efficient between the QD and ligand HOMO and their respective LUMO levels. Disruption of the conjugation that connects the aromatic ring and its substituents to the QD core causes a reduction of the electronic coupling. Our results further support the notion that the ligand/QD complex should be considered as a distinct chemical system with emergent behavior rather than a QD core with ligands whose sole purpose is to passivate surface dangling bonds and prevent agglomeration.
Sigma-Aldrich
Ferrocene, 98%
Sigma-Aldrich
4-Methylbenzenethiol, 98%