Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

539821

Sigma-Aldrich

2-Methylpropane

99.995%

Synonym(s):

Isobutane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CH
CAS Number:
Molecular Weight:
58.12
Beilstein:
1730720
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.01 (21 °C, vs air)

vapor pressure

72.2 psi ( 37.7 °C)

Assay

99.995%

autoignition temp.

860 °F

expl. lim.

8.3 %

bp

−12 °C (lit.)

mp

−160 °C (lit.)

density

2.064 g/mL at 25 °C (lit.)

SMILES string

CC(C)C

InChI

1S/C4H10/c1-4(2)3/h4H,1-3H3

InChI key

NNPPMTNAJDCUHE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Supplied in a carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed.

Compatible with the following:
  • Aldrich® lecture-bottle station systems
  • Aldrich® lecture-bottle gas regulators

Other Notes

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

purge valve

Product No.
Description
Pricing

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-117.4 °F - closed cup

Flash Point(C)

-83 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laure Pison et al.
Physical chemistry chemical physics : PCCP, 17(45), 30328-30342 (2015-10-29)
The solubility of n-butane and 2-methylpropane (isobutane) in three ionic liquids - 1-(2-methylpropyl)-3-methylimidazolium bis(trifluoromethylsulfonyl)imide [(2mC3)C1im][Ntf2], 1-(3-methylbutyl)-3-methylimidazolium bis(trifluoromethylsulfonyl)imide [(3mC4)C1im][Ntf2] and 1-methyl-3-pentylimidazolium bis(trifluoromethylsulfonyl)imide [C5C1im][Ntf2] - has been measured at atmospheric pressure from 303 to 343 K. Isobutane is less soluble than n-butane
Vera Lepperhof et al.
PloS one, 9(9), e107363-e107363 (2014-09-17)
Cell loss after transplantation is a major limitation for cell replacement approaches in regenerative medicine. To assess the survival kinetics of induced pluripotent stem cell (iPSC)-derived cardiomyocytes (CM) we generated transgenic murine iPSC lines which, in addition to CM-specific expression
Richard Wolfenden et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(24), 7484-7488 (2015-06-03)
The hydrophobicities of the 20 common amino acids are reflected in their tendencies to appear in interior positions in globular proteins and in deeply buried positions of membrane proteins. To determine whether these relationships might also have been valid in
Christian Geis et al.
Experimental neurology, 265, 8-21 (2014-12-30)
Neuromyelitis optica (NMO) is causally linked to autoantibodies (ABs) against aquaporin 4 (AQP4). Here, we focused on the pathogenic effects exclusively mediated by human ABs to AQP4 in vivo. We performed cell-free intrathecal (i.th.) passive transfer experiments in Lewis rats
Trieu-Vuong Dinh et al.
Environmental science and pollution research international, 22(12), 9345-9355 (2015-01-21)
The characteristics of volatile organic compounds (VOCs) emitted from several consumer and commercial products (body wash, dishwashing detergent, air freshener, windshield washer fluid, lubricant, hair spray, and insecticide) were studied and compared. The spray products were found to emit the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service