Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

529214

Sigma-Aldrich

2-Fluoro-4-(trifluoromethyl)benzaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F3CC6H3(F)CHO
CAS Number:
Molecular Weight:
192.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.45 (lit.)

bp

118-119 °C (lit.)

density

1.41 g/mL at 25 °C (lit.)

functional group

aldehyde
fluoro

SMILES string

Fc1cc(ccc1C=O)C(F)(F)F

InChI

1S/C8H4F4O/c9-7-3-6(8(10,11)12)2-1-5(7)4-13/h1-4H

InChI key

KFEHNXLFIGPWNB-UHFFFAOYSA-N

Application

2-Fluoro-4-(trifluoromethyl)benzaldehyde may be used in the synthesis of methyl 3-amino-6-(trifluoromethyl)benzo[b]thiophene-2-carboxylate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 548-548 (2014)
Mark C Bagley et al.
Organic & biomolecular chemistry, 13(24), 6814-6824 (2015-05-28)
Microwave irradiation of 2-halobenzonitriles and methyl thioglycolate in the presence of triethylamine in DMSO at 130 °C provides rapid access to 3-aminobenzo[b]thiophenes in 58-96% yield. This transformation has been applied in the synthesis of the thieno[2,3-b]pyridine core motif of LIMK1
Yao Zong et al.
Journal of fluorine chemistry, 203, 193-199 (2018-01-10)
Combretastatin A-1 (CA-1) and combretastatin A-4 (CA-4) isolated from the African bush willow

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service