Skip to Content
MilliporeSigma
All Photos(1)

Documents

527556

Sigma-Aldrich

4-Cyanophenylboronic acid pinacol ester

97%

Synonym(s):

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16BNO2
CAS Number:
Molecular Weight:
229.08
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

95-99 °C (lit.)

SMILES string

CC1(C)OB(OC1(C)C)c2ccc(cc2)C#N

InChI

1S/C13H16BNO2/c1-12(2)13(3,4)17-14(16-12)11-7-5-10(9-15)6-8-11/h5-8H,1-4H3

InChI key

HOPDTPGXBZCBNP-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tryfon Zarganes-Tzitzikas et al.
Molecules (Basel, Switzerland), 24(7) (2019-04-17)
Imaging techniques, such as positron emission tomography (PET), represent great progress in the clinical development of drugs and diagnostics. However, the efficient and timely synthesis of appropriately labeled compounds is a largely unsolved problem. Numerous small drug-like molecules with high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service