911887
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl (bromoethyl)carbamate
Synonym(s):
Benzyl boronate tag, Bromoethyl boronic acid pinacol ester nuclear tag, Nucleus-targeting probe building block
About This Item
Recommended Products
form
liquid
storage temp.
2-8°C
Application
related product
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service