Skip to Content
MilliporeSigma
All Photos(2)

Documents

516090

Sigma-Aldrich

4′-Aminobenzanilide

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CONHC6H4NH2
CAS Number:
Molecular Weight:
212.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

127-131 °C (lit.)

SMILES string

Nc1ccc(NC(=O)c2ccccc2)cc1

InChI

1S/C13H12N2O/c14-11-6-8-12(9-7-11)15-13(16)10-4-2-1-3-5-10/h1-9H,14H2,(H,15,16)

InChI key

GTTFJYUWPUKXJH-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Preparation of 4'-Aminobenzanilide.
Spencer CE.
Journal of the American Chemical Society, 65(12), 2470-2471 (1943)
C R Clark et al.
Journal of medicinal chemistry, 28(9), 1259-1262 (1985-09-01)
A series of 4-aminobenzanilides derived from ring-alkylated anilines were prepared and evaluated for anticonvulsant activity. These benzanilides were prepared in the course of studies designed to determine the relationship between the benzamide structure and anticonvulsant effects. The compounds were tested

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service