Skip to Content
MilliporeSigma
All Photos(1)

Documents

499226

Sigma-Aldrich

Benzyl alcohol, polymer-bound

70-90 mesh, extent of labeling: 2.0-3.0 mmol loading, 1 % cross-linked

Synonym(s):

(Hydroxymethyl)polystyrene, Benzyl alcohol polymer-bound, Hydroxymethyl polystyrene, Polystyrene cross-linked with divinylbenzene, hydroxymethylated

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:
NACRES:
NA.22

Quality Level

crosslinking

1 % cross-linked

reaction suitability

reaction type: Boc solid-phase peptide synthesis

extent of labeling

2.0-3.0 mmol loading

particle size

70-90 mesh

functional group

alcohol

storage temp.

2-8°C

SMILES string

OCc1ccccc1.C=Cc2ccccc2.C=Cc3ccc(C=C)cc3

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service