Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

513555

Sigma-Aldrich

Luperox® 224, 2,4-Pentanedione peroxide solution

~34 wt. % in 4-hydroxy-4-methyl-2-pentanone and N-methyl-2-pyrrolidone

Synonym(s):

2,4-Pentanedione peroxide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O6
CAS Number:
Molecular Weight:
230.21
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

reaction suitability

reagent type: oxidant

concentration

~34 wt. % in 4-hydroxy-4-methyl-2-pentanone and N-methyl-2-pyrrolidone

refractive index

n20/D 1.439

bp

68 °C

density

1.068 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(=O)C(OOC(C(C)=O)C(C)=O)C(C)=O

InChI

1S/C10H14O6/c1-5(11)9(6(2)12)15-16-10(7(3)13)8(4)14/h9-10H,1-4H3

InChI key

ILENATNBFPBMHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Polymerization initiator

Legal Information

Product of Arkema Inc.
Luperox is a registered trademark of Arkema Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Org. Perox. D - Repr. 1B - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 1

Flash Point(F)

214.0 °F - closed cup

Flash Point(C)

101.11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service