Skip to Content
MilliporeSigma
All Photos(1)

Documents

495182

Sigma-Aldrich

Titanium(III) sulfate solution

~45 wt. % in dilute sulfuric acid, ≥99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ti2(SO4)3
CAS Number:
Molecular Weight:
383.92
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.9% trace metals basis

reaction suitability

core: titanium
reagent type: catalyst

concentration

~45 wt. % in dilute sulfuric acid

density

1.456 g/mL at 25 °C

SMILES string

[Ti+3].[Ti+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/3H2O4S.2Ti/c3*1-5(2,3)4;;/h3*(H2,1,2,3,4);;/q;;;2*+3/p-6

InChI key

SOBXOQKKUVQETK-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Used for the synthesis and crystal structure studies of new acid titanium sulfates such as Ti(H5O2)(SO4)2(H2O)2.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saeid Ahmadzadeh et al.
Environmental monitoring and assessment, 190(6), 340-340 (2018-05-12)
Since 2,4-dichlorophenol (2,4-DCP) as a priority pollutant is used in numerous industrial processes, its removal from the aqueous environment is of utmost importance and desire. Herein, the authors describe an electrochemical treatment process for efficient removal of 2,4-DCP from aqueous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service