Skip to Content
MilliporeSigma
All Photos(1)

Documents

89532

Millipore

Titanium(IV) oxysulfate - sulfuric acid solution

27-31% H2SO4 basis, for the detection of H2O2 and ether peroxides

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiOSO4 · (H2SO4)x
CAS Number:
Molecular Weight:
159.93 (free base basis)
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.85

form

liquid

quality

for the detection of H2O2 and ether peroxides

composition

H2SO4, 27-31%
Ti, ~5%

reaction suitability

reagent type: catalyst
core: titanium

density

1.32 g/mL at 20 °C

SMILES string

O=S(O[Ti]1=O)(O1)=O.O=S(O)(O)=O

InChI

1S/2H2O4S.O.Ti/c2*1-5(2,3)4;;/h2*(H2,1,2,3,4);;/q;;;+2/p-2

InChI key

DLRWTNIGAIBGCM-UHFFFAOYSA-L

Application

Titanium(IV) oxysulfate - sulfuric acid solution has been used as solvent for preparing Ti(IV) stock solutions to understand the interaction of oxocarbonic anions with highly charged metal ions using Resonance Raman spectroscopy.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evidences for a localized chromophore in the Ti (IV)/squarate complex: a resonance Raman investigation.
Santos, P. S., et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 56.8, 1553-1562 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service