Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

493929

Sigma-Aldrich

Tripyrrolidinophosphine

97%

Synonym(s):

Phosphorous acid tripyrrolidide, Tris(N,N-tetramethylene)phosphorous acid triamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H24N3P
CAS Number:
Molecular Weight:
241.31
Beilstein:
1343311
MDL number:
UNSPSC Code:
12350000

Assay

97%

refractive index

n20/D 1.53 (lit.)

bp

104 °C/0.1 mmHg (lit.)

density

1.049 g/mL at 25 °C (lit.)

SMILES string

C1CCN(C1)P(N2CCCC2)N3CCCC3

InChI

1S/C12H24N3P/c1-2-8-13(7-1)16(14-9-3-4-10-14)15-11-5-6-12-15/h1-12H2

InChI key

PXFLCAQHOZXYED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service