Skip to Content
MilliporeSigma
All Photos(1)

Documents

490466

Sigma-Aldrich

Oxalic acid-13C2 dihydrate

99 atom % 13C

Synonym(s):

Ethanedioic-13C2 acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO213C13CO2H · 2H2O
CAS Number:
Molecular Weight:
128.05
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

solid

mp

101-102 °C (lit.)

mass shift

M+2

SMILES string

O.O.O[13C](=O)[13C](O)=O

InChI

1S/C2H2O4.2H2O/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);2*1H2/i1+1,2+1;;

InChI key

GEVPUGOOGXGPIO-BQTCFENJSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service