Skip to Content
MilliporeSigma
All Photos(1)

Documents

485381

Sigma-Aldrich

Ammonium nitrate-15N

60 atom % 15N

Synonym(s):

15N ammonium salt, 15N labeled ammonium nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH415NO3
CAS Number:
Molecular Weight:
81.04
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

60 atom % 15N

form

solid

mol wt

80.64 by atom % calculation (60 atom % 15N)

bp

210 °C (lit.)

mp

169 °C (lit.)

application(s)

agriculture

mass shift

M+1

SMILES string

O[N+]([O-])=O.[H][N+]([H])([H])[H]

InChI

1S/HNO3.H3N/c2-1(3)4;/h(H,2,3,4);1H3/i1+1;

InChI key

PRORZGWHZXZQMV-YTBWXGASSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1C - Ammonium nitrate and ammonium nitrate containing preparations

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service