Skip to Content
MilliporeSigma
All Photos(1)

Documents

493295

Sigma-Aldrich

Potassium nitrate-15N,18O3

95 atom % 18O, 98 atom % 15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K15N18O3
CAS Number:
Molecular Weight:
108.10
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % 15N
95 atom % 18O

form

solid

mp

334 °C (lit.)

mass shift

M+7

SMILES string

[K+].[18O-][15N+]([18O-])=[18O]

InChI

1S/K.NO3/c;2-1(3)4/q+1;-1/i;1+1,2+2,3+2,4+2

InChI key

FGIUAXJPYTZDNR-QKBJONFOSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame over circle

Signal Word

Warning

Hazard Statements

Hazard Classifications

Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service