Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

477281

Sigma-Aldrich

(R)-(+)-1,2-Epoxy-9-decene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]25/D +13°, neat

bp

39.5-40.5 °C/0.3 mmHg (lit.)

density

0.86 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CCCCCCC[C@@H]1CO1

InChI

1S/C10H18O/c1-2-3-4-5-6-7-8-10-9-11-10/h2,10H,1,3-9H2/t10-/m1/s1

InChI key

FCZHJHKCOZGQJZ-SNVBAGLBSA-N

General description

(R)-1,2-Epoxy-9-decene is a chiral epoxide used to prepare 4-substituted oxetane-2-ones.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids not in Storage Class 3

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An effective protocol for the synthesis enantiomerically pure 4-substituted oxetane-2-ones
Balakier T, et al.
Tetrahedron, 69(24), 4990-4993 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service