Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

260258

Sigma-Aldrich

1,2-Epoxyoctane

96%

Synonym(s):

1-Octene oxide, Hexyloxirane

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$233.00
100 G
$674.00

$233.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$233.00
100 G
$674.00

About This Item

Empirical Formula (Hill Notation):
C8H16O
CAS Number:
Molecular Weight:
128.21
Beilstein:
79912
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$233.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.42 (lit.)

bp

62.5-63 °C/17 mmHg (lit.)

density

0.839 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCC1CO1

InChI

1S/C8H16O/c1-2-3-4-5-6-8-7-9-8/h8H,2-7H2,1H3

InChI key

NJWSNNWLBMSXQR-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Levin et al.
Archives of biochemistry and biophysics, 220(2), 485-494 (1983-02-01)
Metabolism of cholesterol 5,6 alpha-oxide to the 5,6-glycol is catalyzed by a rat liver microsomal epoxide hydrolase that is distinct from the microsomal epoxide hydrolase that metabolizes a wide range of xenobiotic alkene and arene oxides. The two enzymes are
Jonatan Perez-Arce et al.
Polymers, 13(4) (2021-03-07)
Lignin-based polyols (LBPs) with controlled microstructure were obtained by cationic ring opening polymerization (CROP) of oxiranes in an organosolv lignin (OL) tetrahydrofuran (THF) solution. The control on the microstructure and consequently on the properties of the LBPs such as hydroxyl
Kelli L Hvorecny et al.
Structure (London, England : 1993), 25(5), 697-707 (2017-04-11)
Pseudomonas aeruginosa secretes an epoxide hydrolase with catalytic activity that triggers degradation of the cystic fibrosis transmembrane conductance regulator (CFTR) and perturbs other host defense networks. Targets of this CFTR inhibitory factor (Cif) are largely unknown, but include an epoxy-fatty
Takashi Toyao et al.
ChemSusChem, 8(22), 3905-3912 (2015-09-24)
Various N-doped nanoporous carbons containing metal species were prepared by direct thermal conversion of zeolitic imidazolate frameworks (ZIFs; ZIF-7, -8, -9, and -67) at different temperatures (600, 800, and 1000 °C). These materials were utilized as bifunctional acid-base catalysts to promote

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service