Skip to Content
MilliporeSigma
All Photos(1)

Documents

472964

Sigma-Aldrich

N-Z-D-serine methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(NHCO2CH2C6H5)CO2CH3
CAS Number:
Molecular Weight:
253.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

optical activity

[α]25/D +15°, c = 1 in methanol

reaction suitability

reaction type: solution phase peptide synthesis

bp

170 °C/0.01 mmHg (lit.)

mp

41-43 °C (lit.)

application(s)

peptide synthesis

SMILES string

COC(=O)[C@@H](CO)NC(=O)OCc1ccccc1

InChI

1S/C12H15NO5/c1-17-11(15)10(7-14)13-12(16)18-8-9-5-3-2-4-6-9/h2-6,10,14H,7-8H2,1H3,(H,13,16)/t10-/m1/s1

InChI key

CINAUOAOVQPWIB-SNVBAGLBSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service