Skip to Content
MilliporeSigma
All Photos(3)

Documents

410489

Sigma-Aldrich

Boc-Ser-OMe

95%

Synonym(s):

N-(tert-Butoxycarbonyl)-L-serine methyl ester, Boc-L-serine methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH[NHCO2C(CH3)3]CO2CH3
CAS Number:
Molecular Weight:
219.23
Beilstein:
3545389
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

optical activity

[α]20/D −18°, c = 5 in methanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

refractive index

n20/D 1.452 (lit.)

density

1.082 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

SMILES string

COC(=O)[C@H](CO)NC(=O)OC(C)(C)C

InChI

1S/C9H17NO5/c1-9(2,3)15-8(13)10-6(5-11)7(12)14-4/h6,11H,5H2,1-4H3,(H,10,13)/t6-/m0/s1

InChI key

SANNKFASHWONFD-LURJTMIESA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Griet Van Zeebroeck et al.
Nature chemical biology, 5(1), 45-52 (2008-12-09)
Transporter-related nutrient sensors, called transceptors, mediate nutrient activation of signaling pathways through the plasma membrane. The mechanism of action of transporting and nontransporting transceptors is unknown. We have screened 319 amino acid analogs to identify compounds that act on Gap1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service