Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

470694

Sigma-Aldrich

Methyl 2,6-dimethoxybenzoate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CO2CH3
CAS Number:
Molecular Weight:
196.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

87-90 °C (lit.)

functional group

ester

SMILES string

COC(=O)c1c(OC)cccc1OC

InChI

1S/C10H12O4/c1-12-7-5-4-6-8(13-2)9(7)10(11)14-3/h4-6H,1-3H3

InChI key

XLXVNKPXOIYLLE-UHFFFAOYSA-N

General description

Methyl 2,6-dimethoxybenzoate is a useful inter­mediate for the synthesis of biologically active heterocyclic compounds. Its crystal structure has been investigated. Its molecule has a planar ester group which is at a dihedral angle of 81.46(3)° with respect to the benzene ring.[1]

Application

Methyl 2,6-dimethoxybenzoate may be employed as starting reagent in the total synthesis of (±)-hyperforin.[2]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective total synthesis of (?)-hyperforin via intramolecular cyclopropanation.
Uwamori M and Nakada M.
Tetrahedron Letters, 54(15), 2022-2025 (2013)
Methyl 2, 6-dimethoxybenzoate.
Qadeer G, et al.
Acta Crystallographica Section E, Structure Reports Online, 63(6), 3027-3027 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service