Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

659290

Sigma-Aldrich

Methyl 2-bromo-5-methoxybenzoate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$56.50
25 G
$211.00

$56.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$56.50
25 G
$211.00

About This Item

Linear Formula:
BrC6H3(OCH3)CO2CH3
CAS Number:
Molecular Weight:
245.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$56.50


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.563

bp

271 °C

density

1.520 g/mL at 25 °C

functional group

bromo
ester

SMILES string

COC(=O)c1cc(OC)ccc1Br

InChI

1S/C9H9BrO3/c1-12-6-3-4-8(10)7(5-6)9(11)13-2/h3-5H,1-2H3

InChI key

VRTQLDFCPNVQNT-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service