Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

457639

Sigma-Aldrich

Poly(D,L-lactide-co-caprolactone)

DL-lactide 40 mol %

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

solid

feed ratio

lactide:caprolactone 40:60

composition

DL-lactide, 40 mol %

inherent viscosity

0.7-0.9 dL/g, in chloroform(lit.)

transition temp

Tm (DSC) 31 °C (onset)

storage temp.

2-8°C

SMILES string

CC(O)C(O)=O.OCCCCC(O)=O

InChI key

LLHKFIFJPCHEOM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Poly(DL-lactide-co-caprolactone) may be used to prepare a composite scaffold, which may be used in the development of a functional cardiac tissue.[1]

Features and Benefits

Biocompatible, biodegradable polymer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel composite scaffold for cardiac tissue engineering.
Park H, et al.
In Vitro Cellular & Developmental Biology. Animal, 41(7), 188-196 (2005)

Articles

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

In the past two decades, tissue engineering and regenerative medicine have become important interdisciplinary fields that span biology, chemistry, engineering, and medicine.

Innovations in polymer technology have had a significant impact on the advancement of novel drug delivery systems.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service