Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

453218

Sigma-Aldrich

(3aS,4R,5S,6aR)-(+)-Hexahydro-5-hydroxy-4-(hydroxymethyl)-2H-cyclopenta[b]furan-2-one

98%

Synonym(s):

(+)-Corey lactone

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$375.00

$375.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$375.00

About This Item

Empirical Formula (Hill Notation):
C8H12O4
CAS Number:
Molecular Weight:
172.18
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$375.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

98%

form

solid

optical activity

[α]20/D +44°, c = 2.3 in methanol

mp

114-118 °C (lit.)

functional group

ester
hydroxyl

SMILES string

OC[C@@H]1[C@@H](O)C[C@H]2OC(=O)C[C@@H]12

InChI

1S/C8H12O4/c9-3-5-4-1-8(11)12-7(4)2-6(5)10/h4-7,9-10H,1-3H2/t4-,5-,6-,7+/m0/s1

InChI key

VYTZWRCSPHQSFX-ZTYPAOSTSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service