Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

435643

Sigma-Aldrich

Ethyltrimethoxysilane

≥97%

Synonym(s):

Dow Corning® product 1-6321

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5Si(OCH3)3
CAS Number:
Molecular Weight:
150.25
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

≥97%

form

liquid

refractive index

n20/D 1.384 (lit.)

bp

123-124 °C (lit.)

density

0.94 g/mL at 25 °C (lit.)

SMILES string

CC[Si](OC)(OC)OC

InChI

1S/C5H14O3Si/c1-5-9(6-2,7-3)8-4/h5H2,1-4H3

InChI key

SBRXLTRZCJVAPH-UHFFFAOYSA-N

Legal Information

Repackaged by Aldrich Chemical Company, Inc. Manufactured by Dow Corning Corporation, U.S.A.
Dow Corning is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable Liquids

WGK

WGK 3

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Di Zhou et al.
Journal of chromatography. A, 1627, 461423-461423 (2020-08-22)
A novel stationary phase co-modified with N-isopropyl acrylamide (NIPAM) and 3-aminophenylboronic acid copolymer on the silica was synthesized through atom transfer radical polymerization (ATRP) reaction for performing mixed-mode and boronate affinity chromatography. The prepared functionalized silica was characterized using Fourier
Kellie I McConnell et al.
Journal of controlled release : official journal of the Controlled Release Society, 194, 113-121 (2014-09-03)
There is an unmet clinical need to increase lung transplant successes, patient satisfaction and to improve mortality rates. We offer the development of a nanovector-based solution that will reduce the incidence of lung ischemic reperfusion injury (IRI) leading to graft

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service