Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

427462

Sigma-Aldrich

5-Benzyloxy-1-pentanol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2O(CH2)5OH
CAS Number:
Molecular Weight:
194.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.505 (lit.)

bp

140 °C/0.4 mmHg (lit.)

density

1.008 g/mL at 25 °C (lit.)

SMILES string

OCCCCCOCc1ccccc1

InChI

1S/C12H18O2/c13-9-5-2-6-10-14-11-12-7-3-1-4-8-12/h1,3-4,7-8,13H,2,5-6,9-11H2

InChI key

RZVDPWSEPVHOPU-UHFFFAOYSA-N

General description

5-Benzyloxy-1-pentanol (5-(benzyloxy)pentan-1-ol) has been synthesized by reacting pentane-1,5-diol with benzyl chloride. Its enthalpy of vaporization at boiling point has been determined.

Application

5-Benzyloxy-1-pentanol (5-(benzyloxy)pentan-1-ol) may be used as a starting material in the synthesis of 5-benzyloxypentanal and (±)tanikolide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 514-514 (2008)
Total synthesis of (?) tanikolide.
Zhang R, et al.
Synthetic Communications, 32(14), 2187-2194 (2002)
Ana R Hornillo-Araujo et al.
Organic & biomolecular chemistry, 4(9), 1723-1729 (2006-04-25)
The syntheses of novel tricyclic pyrrolo[2,3-d]pyrimidine analogues of S6-methylthioguanine are described. The crystal structures and pKa values of these and related O6-methylguanine analogues are reported. All compounds display higher pKa values than O6-methylguanine with the sulfur-containing analogues being the more

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service