Skip to Content
MilliporeSigma
All Photos(2)

Documents

459690

Sigma-Aldrich

4-Benzyloxy-1-butanol

97%

Synonym(s):

Tetramethylene glycol monobenzyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2O(CH2)4OH
CAS Number:
Molecular Weight:
180.24
Beilstein:
1944253
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.513 (lit.)

bp

216-256 °C (lit.)

density

1.025 g/mL at 25 °C (lit.)

SMILES string

OCCCCOCc1ccccc1

InChI

1S/C11H16O2/c12-8-4-5-9-13-10-11-6-2-1-3-7-11/h1-3,6-7,12H,4-5,8-10H2

InChI key

TYROJDFHUXSBHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Benzyloxy-1-butanol can be prepared starting from butane-1,4-diol.

Application

4-Benzyloxy-1-butanol may be used for the preparation of ethyl 6-benzyloxy-2-hexenoate, via Swern oxidation and Wadsworth-Emmons olefination. It may be used as starting reagent for the synthesis of 4-(benzyloxy)butanal.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The syntheses of tricyclic analogues of O 6-methylguanine.
Hammond DM, et al.
Organic & Biomolecular Chemistry, 1(23), 4166-4172 (2003)
Improved preparation of ?-hydroxy-a-amino acids: direct formation of sulfates by sulfuryl chloride.
Alonso M and Riera M.
Tetrahedron Asymmetry, 16(23), 3908-3912 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service