Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41304

Sigma-Aldrich

(R)-α-Methyl-2-pyridinemethanol

≥98.0% (GC)

Synonym(s):

(R)-1-(2-Pyridyl)-ethanol, (R)-2-(1-Hydroxyethyl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9NO
CAS Number:
Molecular Weight:
123.15
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

≥98.0% (GC)

form

solid

optical activity

[α]/D +62.0±2.0°, c = 1 in ethanol

optical purity

enantiomeric ratio: ≥99.0:1.0 (GC)

refractive index

n20/D 1.528

storage temp.

2-8°C

SMILES string

C[C@@H](O)c1ccccn1

InChI

1S/C7H9NO/c1-6(9)7-4-2-3-5-8-7/h2-6,9H,1H3/t6-/m1/s1

InChI key

PPHIIIRFJKDTLG-ZCFIWIBFSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service