Skip to Content
MilliporeSigma
All Photos(1)

Documents

ABS2210

Sigma-Aldrich

Anti-ANO6/TMEM16F

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified antibody
serum

antibody product type

primary antibodies

clone

polyclonal

mol wt

calculated mol wt 55.73 and 106.26 kDa
observed mol wt ~58 and 110 kDa

purified by

(Unpurified)

species reactivity

mouse

packaging

antibody small pack of 100 μL

technique(s)

western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

General description

Anoctamin-6 (UniProt: Q6P9J9; also known as Small-conductance calcium-activated nonselective cation channel, SCAN channel, Transmembrane protein 16F) is encoded by the Ano6 (also known as Tmem16f) gene (Gene ID: 105722) in murine species. ANO6/TMEM16F is a multi-pass membrane glycoprotein with six cytoplasmic domains, ten transmembrane domains, and four extracellular domains. It is predominant expressed in epithelial tissues and is also found in skeletal system where it is primarily expressed in osteoblasts. It serves as a small-conductance calcium-activated nonselective cation (SCAN) channel that acts as a regulator of phospholipid scrambling in platelets, osteoblasts and fetal thymocytes. Phospholipid scrambling results in surface exposure of phosphatidylserine which in platelets is essential to trigger the clotting system. In osteoblasts it is essential for the deposition of hydroxyapatite during bone mineralization. ANO6/TMEM16F displays calcium-dependent phospholipid scramblase activity whereby it scrambles phosphatidylserine, phosphatidylcholine, and galactosylceramide. It is shown to be a key component of the plasma membrane repair mechanism triggered by pore-forming agents, such as bacterial toxins. This is achieved by activation of ANO6/TMEM16F by an increase in intracellular calcium resulting from flux of extracellular calcium ion. ANO6/TMEM16F is also shown to generate outwardly rectifying chloride channel currents in airway epithelial cells and Jurkat cells . (Ref.: Wu, N., et al. (2020). Cell Rep. 30(4); 1129-1140).

Specificity

This rabbit polyclonal antibody detects Anoctamin-6 (TMEM16F) in murine species. It targets and epitope within the C-terminal region and detects both isoforms 1 and 2.

Immunogen

GST-tagged recombinant fragment corresponding to 65 amino acids from the C-terminal region of murine Anoctamin-6 (TMEM16F).

Application

Quality Control Testing

Evaluated by Western Blotting in Mouse thymus tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected ANO6/TMEM16F in Mouse thymus tissue lysate.

Tested Applicaitons

Western Blotting Analysis: A representative lot detected ANO6/TMEM16F in Western Blotting applications (Wu, N., et. al. (2020). Cell Rep. 30(4):1129-1140.e5).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-ANO6/TMEM16F, Cat. No. ABS2210, is a rabbit polyclonal antibody that detects Anoctamin-6 and is tested for use in Western Blotting.

Physical form

Rabbit polyclonal antibody in serum with 0.05% sodium azide.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service