Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

410772

Sigma-Aldrich

3-Oxo-1-indancarboxylic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O3
CAS Number:
Molecular Weight:
176.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

111-113 °C (lit.)

SMILES string

OC(=O)C1CC(=O)c2ccccc12

InChI

1S/C10H8O3/c11-9-5-8(10(12)13)6-3-1-2-4-7(6)9/h1-4,8H,5H2,(H,12,13)

InChI key

HXLJFMRZKCSTQD-UHFFFAOYSA-N

General description

3-Oxo-1-indancarboxylic acid participates in the synthesis of 3-hydroxymethyl-1-indanol (±). Enantiomeric separation of 3-oxo-1-indancarboxylic acid on liquid chromatography chiral stationary phases (CSPs) by supercritical fluid chromatography (SFC) has been reported. Separation of 3-oxo-1-indancarboxylic acid enantiomers using a new immobilized polysaccharide chiral stationary phase, CHIRALPAK IA with hexane has been described.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extending the Range of Solvents for Chiral Analysis Using a New Immobilized Polysaccharide Chiral Stationary Phase, CHIRALPAK IA.
Cox GB and Amoss CW.
LCGC North America, 22(2), 32-32 (2004)
Kinetic resolution of 3-hydroxymethylbenzocycloalkanols by selective asymmetric hydrogen-transfer oxidation.
Caro Y, et al.
Tetrahedron Asymmetry, 14(23), 3689-3696 (2003)
Super/subcritical fluid chromatography separations with four synthetic polymeric chiral stationary phases.
Han X, et al.
Chromatographia, 65(7-8), 381-400 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service