Skip to Content
MilliporeSigma
All Photos(1)

Documents

408611

Sigma-Aldrich

Fmoc-Leu-OH

98%

Synonym(s):

N-(9-Fluorenylmethoxycarbonyl)-L-leucine, Fmoc-L-leucine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23NO4
CAS Number:
Molecular Weight:
353.41
Beilstein:
2178254
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

98%

optical activity

[α]20/D −25°, c = 0.5 in DMF

mp

152-156 °C (lit.)

SMILES string

CC(C)C[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C21H23NO4/c1-13(2)11-19(20(23)24)22-21(25)26-12-18-16-9-5-3-7-14(16)15-8-4-6-10-17(15)18/h3-10,13,18-19H,11-12H2,1-2H3,(H,22,25)(H,23,24)/t19-/m0/s1

InChI key

CBPJQFCAFFNICX-IBGZPJMESA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

PPARγ ligand that induces insulin sensitization, but not adipogenesis.

Other Notes

This product has been replaced by 47633-ALDRICH | Fmoc-Leu-OH ≥97.0%

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service