Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

399205

Sigma-Aldrich

trans,trans-Farnesyl chloride

97%

Synonym(s):

trans,trans-1-Chloro-3,7,11-trimethyl-2,6,10-dodecatriene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$134.00
5 G
$460.00

$134.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$134.00
5 G
$460.00

About This Item

Linear Formula:
(CH3)2C=CH[CH2CH2C(CH3)=CH]2CH2Cl
CAS Number:
Molecular Weight:
240.81
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$134.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.493 (lit.)

bp

59-60 °C/2.4 mmHg (lit.)

density

0.916 g/mL at 25 °C (lit.)

functional group

alkyl halide
chloro

storage temp.

2-8°C

SMILES string

[H]\C(CCl)=C(\C)CC\C([H])=C(/C)CC\C([H])=C(/C)C

InChI

1S/C15H25Cl/c1-13(2)7-5-8-14(3)9-6-10-15(4)11-12-16/h7,9,11H,5-6,8,10,12H2,1-4H3/b14-9+,15-11+

InChI key

BJVUJIDTICYHLL-YFVJMOTDSA-N

General description

Physical properties (density, refractive index and boiling point) of trans,trans-farnesyl chloride have been reported.[1]

Application

trans,trans-Farnesyl chloride (all-trans-farnesyl chloride) is suitable for use in the preparation of (S)-farnesyl thiopyrophosphate.[2]
Farnesyl chloride may be used in the synthesis of isoprenoid conjugates of nucleoside 5′-diphosphates.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 487-487 (2015)
Youngha Ryu et al.
Organic letters, 5(24), 4713-4715 (2003-11-25)
[reaction: see text] Isoprenoid conjugates of nucleoside 5'-diphosphates were efficiently synthesized by one-step nucleophilic displacement reactions of either isoprenyl chlorides or isopentenyl tosylate with nucleoside 5'-diphosphates.
Yi-Hung Chen et al.
The Journal of biological chemistry, 277(9), 7369-7376 (2001-12-18)
Undecaprenyl pyrophosphate synthase (UPPS) catalyzes the consecutive condensation reactions of eight isopentenyl pyrophosphate (IPP) with farnesyl pyrophosphate (FPP) to generate C(55) undecaprenyl pyrophosphate (UPP). In the present study, site-directed mutagenesis, fluorescence quenching, and stopped-flow methods were utilized to examine the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service