MilliporeSigma
  • Efficient one-step syntheses of isoprenoid conjugates of nucleoside 5'-diphosphates.
Efficient one-step syntheses of isoprenoid conjugates of nucleoside 5'-diphosphates.
Organic letters (2003-11-25)
Youngha Ryu, A Ian Scott
ABSTRACT
[reaction: see text] Isoprenoid conjugates of nucleoside 5'-diphosphates were efficiently synthesized by one-step nucleophilic displacement reactions of either isoprenyl chlorides or isopentenyl tosylate with nucleoside 5'-diphosphates.
Sigma-Aldrich
trans,trans-Farnesyl chloride, 97%