Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

39736

Sigma-Aldrich

1-Ethyl-3-methylimidazolium tetrafluoroborate

for catalysis, ≥98.5% (T)

Synonym(s):

EMIMBF4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11BF4N2
CAS Number:
Molecular Weight:
197.97
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

for catalysis

Assay

≥98.5% (T)

form

liquid

impurities

≤0.02% water

refractive index

n20/D 1.41
n20/D 1.413 (lit.)

bp

>350 °C (lit.)

mp

15 °C (lit.)

density

1.294 g/mL at 25 °C (lit.)

anion traces

bromide (Br-): ≤10 mg/kg
chloride (Cl-): ≤10 mg/kg
nitrate (NO3-): ≤10 mg/kg
phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤10 mg/kg

SMILES string

F[B-](F)(F)F.CCn1cc[n+](C)c1

InChI

1S/C6H11N2.BF4/c1-3-8-5-4-7(2)6-8;2-1(3,4)5/h4-6H,3H2,1-2H3;/q+1;-1

InChI key

CUNYTRQQXKCRTJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sungwon Lee et al.
Nature communications, 5, 5898-5898 (2014-12-20)
To measure electrophysiological signals from the human body, it is essential to establish stable, gentle and nonallergic contacts between the targeted biological tissue and the electrical probes. However, it is difficult to form a stable interface between the two for
K Kinoshita et al.
Physical chemistry chemical physics : PCCP, 17(28), 18705-18709 (2015-06-30)
The influence of the structure of ionic liquids on the crystallinity of aluminum hydroxide (Al(OH)3) prepared by a sol-gel process with aluminum isopropoxide (Al(OPr(i))3) in imidazolium-based ionic liquids was investigated. When Al(OH)3 was prepared in ionic liquids having long alkyl
Daisuke Sugioka et al.
Physical chemistry chemical physics : PCCP, 17(19), 13150-13159 (2015-04-29)
Sputtering of noble metals, such as Au, Ag, Pd, and Pt, onto room-temperature ionic liquids (RTILs) enabled the formation of monoparticle films composed of spherical noble metal nanoparticles on the liquid surface only when the RTILs used contained hydroxyl-functionalized cations
M T Ubeda-Torres et al.
Journal of chromatography. A, 1380, 96-103 (2015-01-17)
In reversed-phase liquid chromatography in the absence of additives, cationic basic compounds give rise to broad and asymmetrical peaks as a result of ionic interactions with residual free silanols on silica-based stationary phases. Ionic liquids (ILs), added to the mobile

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service