Skip to Content
MilliporeSigma
1-Ethyl-3-methylimidazolium tetrafluoroborate ≥97.0% (HPLC/T)

1-Ethyl-3-methylimidazolium tetrafluoroborate

1-Ethyl-3-methylimidazolium tetrafluoroborate ≥97.0% (HPLC/T)
Synonym(s):
EMIMBF4
Empirical Formula (Hill Notation):
C6H11BF4N2
CAS Number:
Molecular Weight:
197.97

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade