Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

377023

Sigma-Aldrich

2-Iodopropane-d7

98 atom % D, contains copper as stabilizer

Synonym(s):

Isopropyl-d7 iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3)2CDI
CAS Number:
Molecular Weight:
177.04
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

isotopic purity

98 atom % D

Assay

99% (CP)

form

liquid

contains

copper as stabilizer

technique(s)

NMR: suitable

impurities

≤0.5% water
water

refractive index

n20/D 1.4925 (lit.)

bp

88-90 °C

density

1.774 g/mL at 25 °C (lit.)

mass shift

M+7

storage temp.

2-8°C

SMILES string

[2H]C([2H])([2H])C([2H])(I)C([2H])([2H])[2H]

InChI

1S/C3H7I/c1-3(2)4/h3H,1-2H3/i1D3,2D3,3D

InChI key

FMKOJHQHASLBPH-YYWVXINBSA-N

General description

2-Iodopropane-d7 is 2-iodopropane with all the hydrogen atoms substituted by deuterium. Its synthesis from 2-propanol-d8 has been reported.[1]

Application

2-Iodopropane-d7 may be used in the preparation of deuterium labeled prenyldiphosphate and prenylcysteine derivatives,[2] and 1-(isopentylsulfonyl)benzene-d7.[3]

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thangaiah Subramanian et al.
Journal of labelled compounds & radiopharmaceuticals, 56(8), 370-375 (2013-11-29)
A Wittig reaction employing Li(CD3)2CP(C6H5)3 was used to prepare d6-farnesol and d6-geranylgeraniol. Reductive amination of aniline-2,3,4,5,6-d5 was used to prepare the unnatural isoprenoid analogues d5-anilinogeraniol and d5-anilinofarnesol. All of these deuterated isoprenols were elaborated into their diphosphate and cysteine thioether
Maya Kamao et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 859(2), 192-200 (2007-10-19)
Sensitive quantification method for fat-soluble vitamins in human breast milk by liquid chromatography-tandem mass spectrometry was developed. Vitamins A, D and E were extracted from 10.0 mL of breast milk after saponifying by basic condition. Vitamin K derivatives were extracted
Chemistry of the 2, 5-didehydropyridine biradical: Computational, kinetic, and trapping studies toward drug design.
Hoffner J, et al.
Journal of the American Chemical Society, 120(2), 376-385 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service