MilliporeSigma
  • Quantification of fat-soluble vitamins in human breast milk by liquid chromatography-tandem mass spectrometry.
Quantification of fat-soluble vitamins in human breast milk by liquid chromatography-tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2007-10-19)
Maya Kamao, Naoko Tsugawa, Yoshitomo Suhara, Akimori Wada, Toshiyuki Mori, Kazuo Murata, Riichiro Nishino, Tetsuya Ukita, Kazuhiro Uenishi, Kiyoshi Tanaka, Toshio Okano
ABSTRACT
Sensitive quantification method for fat-soluble vitamins in human breast milk by liquid chromatography-tandem mass spectrometry was developed. Vitamins A, D and E were extracted from 10.0 mL of breast milk after saponifying by basic condition. Vitamin K derivatives were extracted from 3.0 mL of breast milk after lipase treatment. The corresponding stable isotope-labeled compounds were used as internal standards. For the determination of vitamin D compounds, derivatization with a Cookson-type reagent was performed. All fat-soluble vitamins were determined by liquid chromatography-tandem mass spectrometry in the positive ion mode. The detection limits of all analytes were 1-250 pg per 50 microL. The recoveries of fat-soluble vitamins were 91-105%. Inter-assay CV values of each vitamin were 1.9-11.9%. The mean concentrations of retinol, vitamin D3, 25-hydroxyvitamin D3, alpha-tocopherol, phylloquinone and menaquinone-4 were 0.455 microg/mL, 0.088 ng/mL, 0.081 ng/mL, 5.087 microg/mL, 3.771 ng/mL, and 1.795 ng/mL, respectively (n=82). This method makes possible to determine fat-soluble vitamins with a wide range of polarities in human breast milk. The assay may be useful for large-scale studies.
Sigma-Aldrich
2-Iodopropane-d7, 98 atom % D, contains copper as stabilizer