Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

371858

Sigma-Aldrich

6-Methoxy-1,2,3,4-tetrahydro-9H-pyrido[3,4-b]indole hydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N2O · HCl
CAS Number:
Molecular Weight:
238.71
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Assay

97%

mp

288-290 °C (lit.)

SMILES string

Cl[H].COc1ccc2[nH]c3CNCCc3c2c1

InChI

1S/C12H14N2O.ClH/c1-15-8-2-3-11-10(6-8)9-4-5-13-7-12(9)14-11;/h2-3,6,13-14H,4-5,7H2,1H3;1H

InChI key

KRMJEIDSHZEQHJ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benny B Tam et al.
Clinical & experimental optometry, 87(3), 171-174 (2004-06-10)
Pinoline is a pineal indoleamine naturally found in the retina. This study compared the effects of pinoline and vitamin E on the copper (I)-induced retinal lipid peroxidation (LPO). Porcine retinal homogenates were mixed with 120 micro M copper (I) solution.
W A Müller et al.
The International journal of developmental biology, 42(6), 825-828 (1998-09-04)
Support or inhibition of DAG-induced head formation: Hydra magnipapillata can be caused to form ectopic head structures by periodictreatmentwith PKC activators such as diacylglycerol (DAG). This ectopic head formation is supported by an extract from the ovine pineal gland that
Anne-Catherine Durand et al.
Journal of enzyme inhibition and medicinal chemistry, 22(5), 556-562 (2007-11-27)
In order to predict the antioxidant activity of 22 pinoline derivatives (1,2,3,4-tetrahydro-beta-carbolines), two dimensional quantitative-structure activity relationships (2D-QSAR) analysis of 19 hexahydropiridoindoles and 12 flavonoids was realized. Five statistically significant models were obtained from randomly constituted training sets (21 compounds)
R Pähkla et al.
Pharmacology & toxicology, 80(3), 122-126 (1997-03-01)
Pinoline (6-methoxy-1,2,3,4-tetrahydro-beta-carboline) is a naturally occurring compound in the mammalian body which inhibits 5-hydroxytryptamine (5-HT) uptake and exerts antidepressant-like behavioural effects in rats. The present study investigates the effects of pinoline on [3H]citalopram binding to the 5-HT transporter on rat
G Piñol-Ripoll et al.
Neuroscience letters, 405(1-2), 89-93 (2006-07-21)
Nitric oxide (NO) is a physiological neurotransmitter, a mediator of the excitatory neurotransmitter glutamate pathways that regulates several neuroendocrine functions, but excessive NO is toxic by itself and it interacts with superoxide radical (O(2)(-)) to form the peroxynitrite anion (ONOO(-)).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service