MilliporeSigma
Effects of vitamin E and pinoline on retinal lipid peroxidation.
Clinical & experimental optometry (2004-06-10)
Benny B Tam, Andrew W Siu, Andy F Lam, Elsa Y Lee
ABSTRACT
Pinoline is a pineal indoleamine naturally found in the retina. This study compared the effects of pinoline and vitamin E on the copper (I)-induced retinal lipid peroxidation (LPO). Porcine retinal homogenates were mixed with 120 micro M copper (I) solution. The mixtures were co-incubated with various concentrations of pinoline or trolox (water-soluble vitamin E analogue) at 37 degrees Centigrade for 60 minutes. The amounts of malondialdehyde (MDA) and protein were assayed to quantify the LPO. Copper (I) ions significantly increased the MDA concentration in the retinal homogenates (p < 0.0007). Both pinoline and trolox significantly suppressed MDA in a dose-dependent manner (p < 0.0001) and their effects were significantly different (p = 0.004). The concentrations that inhibited 50 per cent of LPO were 0.24 mM and 0.68 mM for pinoline and trolox, respectively. Pinoline suppressed the LPO at a potency of 2.8 times compared with trolox. The results support an anti-oxidative role for pinoline in the retina. Further study is required to characterise the pharmacological potency of pinoline in vivo.