Skip to Content
MilliporeSigma
All Photos(1)

Documents

370452

Sigma-Aldrich

2,2-Diethoxyacetamide

97%

Synonym(s):

Glyoxylic amide diethyl acetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2O)2CHCONH2
CAS Number:
Molecular Weight:
147.17
Beilstein:
1760414
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

76-78 °C (lit.)

SMILES string

CCOC(OCC)C(N)=O

InChI

1S/C6H13NO3/c1-3-9-6(5(7)8)10-4-2/h6H,3-4H2,1-2H3,(H2,7,8)

InChI key

DSKGWFQMJHBHRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2-Diethoxyacetamide participates in the synthesis of pyridine-containing central domain of the amythiamicin group of thiopeptide antibiotics.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark C Bagley et al.
Chemical communications (Cambridge, England), (1)(1), 102-103 (2004-01-23)
The pyridine-containing central domain of the amythiamicin group of thiopeptide antibiotics is prepared in protected form in 9 steps, 93%ee and 18% overall yield from (S)-2-[1-(tert-butoxycarbonylamino)-2-methylpropyl]thiazole-4-carboxylic acid by Michael addition-cyclodehydration of a 2-(2-thiazolyl)enamine and 1-(2-thiazolyl)propynone.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service