Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

369527

Sigma-Aldrich

N,N-Dimethylbutylamine

99%

Synonym(s):

N-Butyldimethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3N(CH3)2
CAS Number:
Molecular Weight:
101.19
Beilstein:
1731308
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

refractive index

n20/D 1.398 (lit.)

bp

93.3 °C/750 mmHg (lit.)

density

0.721 g/mL at 25 °C (lit.)

SMILES string

CCCCN(C)C

InChI

1S/C6H15N/c1-4-5-6-7(2)3/h4-6H2,1-3H3

InChI key

DJEQZVQFEPKLOY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethylbutylamine is a tertiaryalkylamine. X-ray photoelectron spectra (XPS) of Si(001) surfaces exposed to N,N-dimethylbutylamine at 300K has been reported.

Application

N,N-Dimethylbutylamine is suitable for use in the fabrication of polystyrene-based nano-LC monolithic columns for the separation of protein molecules. It may be used as ion-pairing reagent in a study involving comparison of performance of six ion-pairing reagents as mobile phase modifiers for oligonucleotide LC/MS.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

23.0 °F - closed cup

Flash Point(C)

-5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joshua E S J Reid et al.
Physical chemistry chemical physics : PCCP, 19(41), 28133-28138 (2017-10-13)
The ionic nature of a functionalized protic ionic liquid cannot be rationalized simply through the differences in aqueous proton dissociation constants between the acid precursor and the conjugate acid of the base precursor. The extent of proton transfer, i.e. the
Leta Deressa Tolesa et al.
International journal of biological macromolecules, 130, 818-826 (2019-03-07)
Ammonium-based ionic liquids (ILs): diisopropylethylammonium acetate ([DIPEA][Ac]), diisopropylethylammonium propanoate ([DIPEA][P]) and dimethylbutylammonium acetate ([DMBA][Ac]) are used for the extraction of chitin from shrimp shells. The extracted chitins were characterized by FT-IR, TGA, XRD, SEM and 1H NMR. The yield of
Anna Kilanowska et al.
Journal of chromatography. A, 1622, 461100-461100 (2020-05-04)
The aim of the present investigation was application of hydrophilic interaction liquid chromatography as an alternative chromatographic approach for the study of antisense oligonucleotides. The influence of several mobile phases, differing with the salt type, their concentration and pH value
Łukasz Nuckowski et al.
The Analyst, 144(15), 4622-4632 (2019-06-28)
Our research focused on applying microextraction by packed sorbent to extracting antisense oligonucleotides from serum samples. The tested sorbents included poly(styrene-co-divinylbenzene), octyl, octadecyl, and unmodified silica gel. As nonpolar sorbents were used for highly-polar molecules, this required ion-pair mode. Comprehensive
Lingzhi Gong et al.
Rapid communications in mass spectrometry : RCM, 28(4), 339-350 (2014-01-08)
A sensitive and selective liquid chromatography/mass spectrometry (LC/MS) method is essential for quality control of synthetic oligonucleotides. However, researchers are still searching for improvements to ion-pairing reagents for ion-pairing reversed-phase LC/MS. This study performed a comprehensive comparison of six ion-pairing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service