Skip to Content
MilliporeSigma
All Photos(1)

Documents

190187

Sigma-Aldrich

1,2-Dimethylpropylamine

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH(CH3)NH2
CAS Number:
Molecular Weight:
87.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97%

refractive index

n20/D 1.4055 (lit.)

bp

84-87 °C (lit.)

mp

−50 °C (lit.)

density

0.757 g/mL at 25 °C (lit.)

SMILES string

CC(C)C(C)N

InChI

1S/C5H13N/c1-4(2)5(3)6/h4-5H,6H2,1-3H3

InChI key

JOZZAIIGWFLONA-UHFFFAOYSA-N

Application

1,2-Dimethylpropylamine was used as standard in separation of alkali and alkaline earth metal cations by capillary electrochromatography on monolithic octadecylsilica columns.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thanh Duc Mai et al.
Analytica chimica acta, 653(2), 228-233 (2009-10-08)
A fast separation of alkali and alkaline earth metal cations and ammonium was carried out by capillary electrochromatography on monolithic octadecylsilica columns of 15 cm length and 100 microm inner diameter using water/methanol mixtures containing acetic acid as mobile phase.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service