Skip to Content
MilliporeSigma
All Photos(2)

Documents

345725

Sigma-Aldrich

2,3,6-Trifluorophenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F3C6H2OH
CAS Number:
Molecular Weight:
148.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

33-37 °C (lit.)

SMILES string

Oc1c(F)ccc(F)c1F

InChI

1S/C6H3F3O/c7-3-1-2-4(8)6(10)5(3)9/h1-2,10H

InChI key

QSFGUSFDWCVXNR-UHFFFAOYSA-N

General description

2,3,6-Trifluorophenol is a fluorinated phenol. Biotransformation of 2,3,6-trifluorophenol has been studied by 19FNMR technique.

Application

2,3,6-Trifluorophenol may be employed as model compound to investigate the site-selective functionalization of halogen-bearing phenols by organometallic approach.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M G Boersma et al.
Journal of industrial microbiology & biotechnology, 26(1-2), 22-34 (2001-09-11)
Of all NMR-observable isotopes 19F is the one most convenient for studies on the biodegradation of environmental pollutants and especially for fast initial metabolic screening of newly isolated organisms. In the past decade we have identified the 19F NMR characteristics
The site-selective functionalization of halogen-bearing phenols: an exercise in diversity-oriented organometallic synthesis.
Marzi E and Schlosser M.
Tetrahedron, 61(13), 3393-3401 (2005)
Matthew G Romei et al.
Science (New York, N.Y.), 367(6473), 76-79 (2020-01-04)
Rotation around a specific bond after photoexcitation is central to vision and emerging opportunities in optogenetics, super-resolution microscopy, and photoactive molecular devices. Competing roles for steric and electrostatic effects that govern bond-specific photoisomerization have been widely discussed, the latter originating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service