MilliporeSigma
Electrostatic control of photoisomerization pathways in proteins.
Science (New York, N.Y.) (2020-01-04)
Matthew G Romei, Chi-Yun Lin, Irimpan I Mathews, Steven G Boxer
ABSTRACT
Rotation around a specific bond after photoexcitation is central to vision and emerging opportunities in optogenetics, super-resolution microscopy, and photoactive molecular devices. Competing roles for steric and electrostatic effects that govern bond-specific photoisomerization have been widely discussed, the latter originating from chromophore charge transfer upon excitation. We systematically altered the electrostatic properties of the green fluorescent protein chromophore in a photoswitchable variant, Dronpa2, using amber suppression to introduce electron-donating and electron-withdrawing groups to the phenolate ring. Through analysis of the absorption (color), fluorescence quantum yield, and energy barriers to ground- and excited-state isomerization, we evaluate the contributions of sterics and electrostatics quantitatively and demonstrate how electrostatic effects bias the pathway of chromophore photoisomerization, leading to a generalized framework to guide protein design.
Sigma-Aldrich
3-Chloro-L-tyrosine, 97%
Sigma-Aldrich
2,3,6-Trifluorophenol, 98%