Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

336955

Sigma-Aldrich

Tri-tert-butylphosphine

90%, technical grade

Synonym(s):

P(t-Bu)3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]3P
CAS Number:
Molecular Weight:
202.32
Beilstein:
1738613
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

technical grade

Assay

90%

reaction suitability

reaction type: Cross Couplings
reagent type: ligand

bp

102-103 °C/13 mmHg (lit.)

mp

30-35 °C (lit.)

density

0.834 g/mL at 20 °C (lit.)

functional group

phosphine

SMILES string

CC(C)(C)P(C(C)(C)C)C(C)(C)C

InChI

1S/C12H27P/c1-10(2,3)13(11(4,5)6)12(7,8)9/h1-9H3

InChI key

BWHDROKFUHTORW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Pyr. Sol. 1 - Skin Corr. 1B

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minseon Kong et al.
Nanomaterials (Basel, Switzerland), 10(12) (2020-12-19)
The leakage and volatilization of liquid electrolytes limit the commercialization of dye-sensitized solar cells (DSCs). As solid-state (ss) hole-transporting materials, free from leakage and volatilization, biscarbazole-based polymers with different molecular weights (PBCzA-H (21,200 g/mol) and PBCzA-L (2450 g/mol)) were applied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service