Skip to Content
MilliporeSigma
Tri-tert-butylphosphine 98%

Tri-tert-butylphosphine

Tri-tert-butylphosphine 98%
Synonym(s):
P(t-Bu)3
Linear Formula:
[(CH3)3C]3P
CAS Number:
Molecular Weight:
202.32
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing