Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

325589

Sigma-Aldrich

Erbium(III) carbonate hydrate

99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Er2(CO3)3·xH2O
CAS Number:
Molecular Weight:
514.54 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302

Assay

99.9%

SMILES string

O.[Er].OC(O)=O


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service