Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

310336

Sigma-Aldrich

cis-4,7-Dihydro-1,3-dioxepin

97%

Synonym(s):

cis-1,3-Dioxep-5-ene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O2
CAS Number:
Molecular Weight:
100.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.457 (lit.)

bp

125-127 °C (lit.)

density

1.049 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

C1OCC=CCO1

InChI

1S/C5H8O2/c1-2-4-7-5-6-3-1/h1-2H,3-5H2

InChI key

BAKUAUDFCNFLBX-UHFFFAOYSA-N

General description

4,7-dihydro-1,3-dioxepin derivatives undergoes hydroformylation to form optically active aldehydes.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshihide Horiuchi et al.
The Journal of organic chemistry, 62(13), 4285-4292 (1997-06-27)
Asymmetric hydroformylation of heterocyclic olefins catalyzed by phosphine-phosphite-Rh(I) complexes has been investigated. Hydroformylation of symmetrical heterocyclic olefins such as 2,5-dihydrofuran, 3-pyrroline derivatives, and 4,7-dihydro-1,3-dioxepin derivatives afforded the optically active aldehydes as single products in 64-76% ee. Unsymmetrical substrates such as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service