Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D59607

Sigma-Aldrich

1,4-Dichloro-2-butyne

99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$119.00
25 G
$362.00
100 G
$1,250.00

$119.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$119.00
25 G
$362.00
100 G
$1,250.00

About This Item

Linear Formula:
ClCH2C≡CCH2Cl
CAS Number:
Molecular Weight:
122.98
Beilstein:
635740
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$119.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.505 (lit.)

bp

165-168 °C (lit.)

density

1.258 g/mL at 25 °C (lit.)

SMILES string

ClCC#CCCl

InChI

1S/C4H4Cl2/c5-3-1-2-4-6/h3-4H2

InChI key

RCHDLEVSZBOHOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service