Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

297941

Sigma-Aldrich

2-Methoxy-3-methylpyrazine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2O
CAS Number:
Molecular Weight:
124.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

refractive index

n20/D 1.507 (lit.)

density

1.06 g/mL at 25 °C (lit.)

SMILES string

COc1nccnc1C

InChI

1S/C6H8N2O/c1-5-6(9-2)8-4-3-7-5/h3-4H,1-2H3

InChI key

VKJIAEQRKBQLLA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lowest observable adverse effect levels of 2-methoxy-3-methylpyrazine which inhibits ciliary beat frequency, oocyte pickup rate and infundibular smooth muscle contraction in the hamster oviduct has been evaluated.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karen Riveles et al.
Reproductive biology and endocrinology : RB&E, 2, 23-23 (2004-05-14)
Our past studies have shown that cigarette smoke inhibits oviductal functioning in vivo and in vitro. The goals in this study were to identify pyrazine derivatives in cigarette smoke solutions that inhibit ciliary beat frequency, oocyte pickup rate, and infundibular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service